4'-(Methylthio)acetophenone 97%

BD18925-25G BD18925-5G
BLDPBD18925-25GEA 38.9 EUR
BLDPBD18925-25G BLDPBD18925-5G
4'-(Methylthio)acetophenone 97%
4'-(Methylthio)acetophenone

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₁₀OS
Storage Temperature: Ambient
MDL Number: MFCD00039835
CAS Number: 1778-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR