(R)-(-)-2-Oxothiazolidine-4-carboxylic acid 98%

BD19158-10G BD19158-100G
BLDPBD19158-10GEA 65.1 EUR
BLDPBD19158-10G BLDPBD19158-100G
(R)-(-)-2-Oxothiazolidine-4-carboxylic acid 98%
(R)-(-)-2-Oxothiazolidine-4-carboxylic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₄H₅NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00066092
CAS Number: 19771-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR