Fmoc-2-methyl-L-phenylalanine 95%

BD19340-10G BD19340-5G
BLDPBD19340-10GEA 378 EUR
BLDPBD19340-10G BLDPBD19340-5G
Fmoc-2-methyl-L-phenylalanine 95%
Fmoc-2-methyl-L-phenylalanine
Formula: C₂₅H₂₃NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00671391
CAS Number: 211637-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR