2-Amino-4,5-dimethyl-1H-pyrrole-3-carbonitrile 95%
|
|
BD19358-1G
BD19358-25G
BLDPBD19358-1GEA
13.7
EUR
BLDPBD19358-1G
BLDPBD19358-25G
2-Amino-4,5-dimethyl-1H-pyrrole-3-carbonitrile 95%
2-Amino-4,5-dimethyl-1H-pyrrole-3-carbonitrile
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₇H₉N₃ MW: 135.17 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00849263 CAS Number: 21392-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...