Methyl-3,4-dimethoxybenzoate 98%

BD19374-500G BD19374-100G
BLDPBD19374-500GEA 411 EUR
BLDPBD19374-500G BLDPBD19374-100G
Methyl-3,4-dimethoxybenzoate 98%
Methyl 3,4-dimethoxybenzoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00008430
CAS Number: 2150-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR