Boc-Orn-OH 97%

BD19411-10G BD19411-1G
BLDPBD19411-10GEA 43.1 EUR
BLDPBD19411-10G BLDPBD19411-1G
Boc-Orn-OH 97%
Boc-Orn-OH

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₀H₂₀N₂O₄
MW: 232.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00076970
CAS Number: 21887-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR