2-Bromo-4-chloropyridine 97%

BD19514-5G BD19514-1G
BLDPBD19514-5GEA 24.2 EUR
BLDPBD19514-5G BLDPBD19514-1G
2-Bromo-4-chloropyridine 97%
2-Bromo-4-chloropyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₅H₃BrClN
Storage Temperature: Ambient
MDL Number: MFCD03840757
CAS Number: 22918-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR