2-Chloro-6-iodo-3-pyridinol 97%
|
|
BD196502-25G
BD196502-1G
BLDPBD196502-25GEA
239
EUR
BLDPBD196502-25G
BLDPBD196502-1G
2-Chloro-6-iodo-3-pyridinol 97%
2-Chloro-6-iodo-3-pyridinol
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₅H₃ClINO Storage Temperature: Refrigerator |
MDL Number:
MFCD10566651 CAS Number: 185220-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...