S-tert-Butyl-L-cysteine hydrochloride 98%

BD19655-1G BD19655-250MG
BLDPBD19655-1GEA 10.5 EUR
BLDPBD19655-1G BLDPBD19655-250MG
S-tert-Butyl-L-cysteine hydrochloride 98%
S-tert-Butyl-L-cysteine hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formula: C₇H₁₆ClNO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD00037426
CAS Number: 2481-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR