Ethyl-3-amino-1H-pyrrole-2-carboxylate hydrochloride 97%
|
|
BD19701-1G
BD19701-10G
BLDPBD19701-1GEA
17.9
EUR
BLDPBD19701-1G
BLDPBD19701-10G
Ethyl-3-amino-1H-pyrrole-2-carboxylate hydrochloride 97%
Ethyl-3-amino-1H-pyrrole-2-carboxylate hydrochloride
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₇H₁₁ClN₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD04038917 CAS Number: 252932-49-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...