(2R)-3-Bromo-2-hydroxy-2-methylpropanoic acid 97%

BD19795-25G BD19795-1G BD19795-10G
BLDPBD19795-25GEA 225 EUR
BLDPBD19795-25G BLDPBD19795-1G BLDPBD19795-10G
(2R)-3-Bromo-2-hydroxy-2-methylpropanoic acid 97%
(2R)-3-Bromo-2-hydroxy-2-methylpropanoic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₄H₇BrO₃
Storage Temperature: Ambient
MDL Number: MFCD09751022
CAS Number: 261904-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR