trans-Indole-3-acrylic acid 95%

BD20062-25G BD20062-250MG
BLDPBD20062-25GEA 403 EUR
BLDPBD20062-25G BLDPBD20062-250MG
trans-Indole-3-acrylic acid 95%
trans-Indole-3-acrylic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₁H₉NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00005633
CAS Number: 29953-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR