3-Bromo-L-phenylalanine 98%

BD20074-1G BD20074-10G
BLDPBD20074-1GEA 7.4 EUR
BLDPBD20074-1G BLDPBD20074-10G
3-Bromo-L-phenylalanine 98%
3-Bromo-L-phenylalanine

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₁₀BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD06659110
CAS Number: 82311-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR