3,4-Difluoro-L-phenylalanine 97%

BD20137-250MG BD20137-25G
BLDPBD20137-250MGEA 16.8 EUR
BLDPBD20137-250MG BLDPBD20137-25G
3,4-Difluoro-L-phenylalanine 97%
3,4-Difluoro-L-phenylalanine

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₉F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD01075125
CAS Number: 31105-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR