5-Fluoroisatoic anhydride 97%

BD20210-1G BD20210-100G
BLDPBD20210-1GEA 17.9 EUR
BLDPBD20210-1G BLDPBD20210-100G
5-Fluoroisatoic anhydride 97%
5-Fluoroisatoic anhydride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₄FNO₃
Storage Temperature: Ambient
MDL Number: MFCD03300719
CAS Number: 321-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR