(2R,2'R)-Dimethyl-3,3'-disulphanediylbis(2-aminopropanoate) dihydrochloride 97%

BD20244-25G BD20244-100G
BLDPBD20244-25GEA 39.9 EUR
BLDPBD20244-25G BLDPBD20244-100G
(2R,2'R)-Dimethyl-3,3'-disulphanediylbis(2-aminopropanoate) dihydrochloride 97%
(2R,2'R)-Dimethyl-3,3'-disulphanediylbis(2-aminopropanoate) dihydrochloride

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₁₈Cl₂N₂O₄S₂
Storage Temperature: Ambient
MDL Number: MFCD00012490
CAS Number: 32854-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR