Uridine 5′-monophosphate disodium salt 98%

BD20303-25G BD20303-500G
BLDPBD20303-25GEA 17.9 EUR
BLDPBD20303-25G BLDPBD20303-500G
Uridine 5′-monophosphate disodium salt 98%
Uridine 5′-monophosphate disodium salt

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₉H₁₁N₂Na₂O₉P
MW: 368.15 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00006525
CAS Number: 3387-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR