4,4'-Difluorobenzhydrol 98%

BD20466-500G BD20466-25G
BLDPBD20466-500GEA 173 EUR
BLDPBD20466-500G BLDPBD20466-25G
4,4'-Difluorobenzhydrol 98%
4,4'-Difluorobenzhydrol

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₃H₁₀F₂O
MW: 220.22 g/mol
Boiling Pt: 305
Melting Pt: 47
Storage Temperature: Ambient
MDL Number: MFCD00000357
CAS Number: 365-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR