2-(4-Bromophenyl)pyrrolidine 97%
|
|
BD20575-5G
BD20575-250MG
BLDPBD20575-5GEA
268
EUR
BLDPBD20575-5G
BLDPBD20575-250MG
2-(4-Bromophenyl)pyrrolidine 97%
2-(4-Bromophenyl)pyrrolidine
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₁₀H₁₂BrN Storage Temperature: Refrigerator |
MDL Number:
MFCD02663432 CAS Number: 383127-22-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...