N-(2-Hydroxyethyl)phthalimide 98%

BD20595-10G BD20595-500G
BLDPBD20595-10GEA 6.3 EUR
BLDPBD20595-10G BLDPBD20595-500G
N-(2-Hydroxyethyl)phthalimide 98%
N-(2-Hydroxyethyl)phthalimide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₀H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD00005903
CAS Number: 3891-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR