6-Fluoropicolinic acid 98%

BD20672-25G BD20672-5G
BLDPBD20672-25GEA 71.4 EUR
BLDPBD20672-25G BLDPBD20672-5G
6-Fluoropicolinic acid 98%
6-Fluoropicolinic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₆H₄FNO₂
Storage Temperature: Ambient
MDL Number: MFCD02181193
CAS Number: 402-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR