6-Bromo-1-hexanol 97%

BD20806-5G BD20806-10G BD20806-1G
BLDPBD20806-5GEA 7.4 EUR
BLDPBD20806-5G BLDPBD20806-10G BLDPBD20806-1G
6-Bromo-1-hexanol 97%
6-Bromo-1-hexanol

Storage condition: Inert atmosphere, 2-8°C

Formula: BrH₂C(CH₂)₄CH₂OH
MW: 181.07 g/mol
Boiling Pt: 105
Storage Temperature: Refrigerator
MDL Number: MFCD00002983
CAS Number: 4286-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR