Methyl-4-chloro-3-methoxy-5-nitrobenzenecarboxylate 97%

BD208081-25G BD208081-100G
BLDPBD208081-25GEA 195 EUR
BLDPBD208081-25G BLDPBD208081-100G
Methyl-4-chloro-3-methoxy-5-nitrobenzenecarboxylate 97%
Methyl-4-chloro-3-methoxy-5-nitrobenzenecarboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₈ClNO₅
Storage Temperature: Ambient
MDL Number: MFCD16140357
CAS Number: 63603-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR