5-Bromo-1H-pyrrolo[3,4-b]pyridin-3-amine 97%
|
|
BD208713-1G
BD208713-5G
BLDPBD208713-1GEA
324
EUR
BLDPBD208713-1G
BLDPBD208713-5G
5-Bromo-1H-pyrrolo[3,4-b]pyridin-3-amine 97%
5-Bromo-1H-pyrrolo[3,4-b]pyridin-3-amine
Storage condition: Keep in dark place, Sealed in dry, 2-8°C
Formula:
C₇H₆BrN₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD17012797 CAS Number: 507462-51-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...