trans-2,6-Dimethylmorpholine 95%

BD208857-100MG BD208857-250MG
BLDPBD208857-100MGEA 43.1 EUR
BLDPBD208857-100MG BLDPBD208857-250MG
trans-2,6-Dimethylmorpholine 95%
trans-2,6-Dimethylmorpholine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₆H₁₃NO
Storage Temperature: Refrigerator
MDL Number: MFCD09953181
CAS Number: 6485-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR