5-Methyl-N-carbobenzoxy-L-glutamate 98%

BD20919-25G BD20919-250MG
BLDPBD20919-25GEA 118 EUR
BLDPBD20919-25G BLDPBD20919-250MG
5-Methyl-N-carbobenzoxy-L-glutamate 98%
5-Methyl-N-carbobenzoxy-L-glutamate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₄H₁₇NO₆
MW: 295.29 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00191090
CAS Number: 4652-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR