N-Methyl-1,2-phenylenediamine 97%
|
|
BD20945-500G
BD20945-10G
BLDPBD20945-500GEA
941
EUR
BLDPBD20945-500G
BLDPBD20945-10G
N-Methyl-1,2-phenylenediamine 97%
N-Methyl-1,2-phenylenediamine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₇H₁₀N₂ Storage Temperature: Ambient |
MDL Number:
MFCD00192314 CAS Number: 4760-34-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...