Methyl-3-bromo-1H-indazole-6-carboxylate 97%
|
|
BD209901-5G
BD209901-250MG
BLDPBD209901-5GEA
175
EUR
BLDPBD209901-5G
BLDPBD209901-250MG
Methyl-3-bromo-1H-indazole-6-carboxylate 97%
Methyl 3-bromo-1H-indazole-6-carboxylate
Storage condition: Sealed in dry, 2-8°C
Formula:
C₉H₇BrN₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD11499017 CAS Number: 192945-56-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...