(5-Carboxypentyl)triphenylphosphonium bromide 97%
|
|
BD21092-5G
BD21092-25G
BLDPBD21092-5GEA
13.7
EUR
BLDPBD21092-5G
BLDPBD21092-25G
(5-Carboxypentyl)triphenylphosphonium bromide 97%
(5-Carboxypentyl)triphenylphosphonium bromide
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₂₄H₂₆BrO₂P Storage Temperature: Ambient |
MDL Number:
MFCD00055556 CAS Number: 50889-29-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...