1,4-Bis(2-benzoxazolyl)naphthalene 96%
|
|
BD21094-25G
BD21094-1000G
BLDPBD21094-25GEA
15.8
EUR
BLDPBD21094-25G
BLDPBD21094-1000G
1,4-Bis(2-benzoxazolyl)naphthalene 96%
1,4-Bis(2-benzoxazolyl)naphthalene
Storage condition: Sealed in dry, Room Temperature
Formula:
C₂₄H₁₄N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00357168 CAS Number: 5089-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...