2-Chloro-p-anisidine 97%
|
|
BD211302-100G
BD211302-1G
BLDPBD211302-100GEA
616
EUR
BLDPBD211302-100G
BLDPBD211302-1G
2-Chloro-p-anisidine 97%
2-Chloro-p-anisidine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₇H₈ClNO MW: 157.6000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00209639 CAS Number: 29242-84-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...