DL-Tryptophan 98%

BD212306-5G BD212306-100G
BLDPBD212306-5GEA 7.4 EUR
BLDPBD212306-5G BLDPBD212306-100G
DL-Tryptophan 98%
DL-Tryptophan

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₁H₁₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00064339
CAS Number: 54-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR