2-Hydroxy-4-methoxybenzonitrile 97%

BD212355-25G BD212355-1G
BLDPBD212355-25GEA 564 EUR
BLDPBD212355-25G BLDPBD212355-1G
2-Hydroxy-4-methoxybenzonitrile 97%
2-Hydroxy-4-methoxybenzonitrile

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₇NO₂
MW: 149.1500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08567959
CAS Number: 39835-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR