2,5-Difluoro-4-iodopyridine 98%

BD213074-25G BD213074-100G
BLDPBD213074-25GEA 257 EUR
BLDPBD213074-25G BLDPBD213074-100G
2,5-Difluoro-4-iodopyridine 98%
2,5-Difluoro-4-iodopyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₅H₂F₂IN
Storage Temperature: Refrigerator
MDL Number: MFCD11040606
CAS Number: 1017793-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR