Ethyl-3-bromopropionate 98%
|
|
BD21311-100G
BD21311-500G
BLDPBD21311-100GEA
28.4
EUR
BLDPBD21311-100G
BLDPBD21311-500G
Ethyl-3-bromopropionate 98%
Ethyl 3-bromopropionate
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₅H₉BrO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00000251 CAS Number: 539-74-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...