3-Fluoro-2-nitropyridine 98%

BD21336-25G BD21336-10G
BLDPBD21336-25GEA 174 EUR
BLDPBD21336-25G BLDPBD21336-10G
3-Fluoro-2-nitropyridine 98%
3-Fluoro-2-nitropyridine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₅H₃FN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD04114127
CAS Number: 54231-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR