5-Iodo-2'-deoxyuridine (Idoxuridine) 98%

BD21350-25G BD21350-10G
BLDPBD21350-25GEA 170 EUR
BLDPBD21350-25G BLDPBD21350-10G
5-Iodo-2'-deoxyuridine (Idoxuridine) 98%
5-Iodo-2'-deoxyuridine (Idoxuridine)

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₁₁IN₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00134656
CAS Number: 54-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR