Benzo[b]thiophene-2-carbonitrile 95%

BD21415-25G BD21415-5G
BLDPBD21415-25GEA 1010 EUR
BLDPBD21415-25G BLDPBD21415-5G
Benzo[b]thiophene-2-carbonitrile 95%
Benzo[b]thiophene-2-carbonitrile

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₉H₅NS
Storage Temperature: Refrigerator
MDL Number: MFCD09751294
CAS Number: 55219-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR