2-Bromo-5-hydroxypyridine 98%

BD21450-250MG BD21450-5G
BLDPBD21450-250MGEA 6.3 EUR
BLDPBD21450-250MG BLDPBD21450-5G
2-Bromo-5-hydroxypyridine 98%
2-Bromo-5-hydroxypyridine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₅H₄BrNO
Storage Temperature: Ambient
MDL Number: MFCD04114184
CAS Number: 55717-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR