(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid 98%
|
|
BD21500-1G
BD21500-250MG
BD21500-5G
BLDPBD21500-1GEA
74.6
EUR
BLDPBD21500-1G
BLDPBD21500-250MG
BLDPBD21500-5G
(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid 98%
(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid
Storage condition: Sealed in dry, 2-8°C
Formula:
C₁₁H₁₉NO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD01862939 CAS Number: 56502-01-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...