5-Bromo-2,4-dimethoxypyrimidine 98%

BD21514-100G BD21514-10G
BLDPBD21514-100GEA 115 EUR
BLDPBD21514-100G BLDPBD21514-10G
5-Bromo-2,4-dimethoxypyrimidine 98%
5-Bromo-2,4-dimethoxypyrimidine

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₇BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00038016
CAS Number: 56686-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR