Methyl-2,2-difluorobenzo[d][1,3]dioxole-5-carboxylate 98%

BD215235-100G BD215235-25G
BLDPBD215235-100GEA 299 EUR
BLDPBD215235-100G BLDPBD215235-25G
Methyl-2,2-difluorobenzo[d][1,3]dioxole-5-carboxylate 98%
Methyl-2,2-difluorobenzo[d][1,3]dioxole-5-carboxylate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₆F₂O₄
Storage Temperature: Ambient
MDL Number: MFCD06203771
CAS Number: 773873-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR