5,6-Dichloropicolinic acid 98%

BD215239-25G BD215239-250MG
BLDPBD215239-25GEA 699 EUR
BLDPBD215239-25G BLDPBD215239-250MG
5,6-Dichloropicolinic acid 98%
5,6-Dichloropicolinic acid

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₃Cl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD13185546
CAS Number: 88912-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR