2-(4-Diethylamino-2-hydroxybenzoyl)benzoic acid 98%
|
|
BD21589-5G
BD21589-25G
BLDPBD21589-5GEA
7.4
EUR
BLDPBD21589-5G
BLDPBD21589-25G
2-(4-Diethylamino-2-hydroxybenzoyl)benzoic acid 98%
2-(4-Diethylamino-2-hydroxybenzoyl)benzoic acid
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₈H₁₉NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00134669 CAS Number: 5809-23-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...