2,4-Dibromopyridine 98%

BD21610-100G BD21610-5G
BLDPBD21610-100GEA 165 EUR
BLDPBD21610-100G BLDPBD21610-5G
2,4-Dibromopyridine 98%
2,4-Dibromopyridine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₅H₃Br₂N
Storage Temperature: Ambient
MDL Number: MFCD01859720
CAS Number: 58530-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR