(S)-1-(2,4-Difluorophenyl)ethanamine hydrochloride 98%
|
|
BD216347-250MG
BD216347-1G
BLDPBD216347-250MGEA
96.6
EUR
BLDPBD216347-250MG
BLDPBD216347-1G
(S)-1-(2,4-Difluorophenyl)ethanamine hydrochloride 98%
(S)-1-(2,4-Difluorophenyl)ethanamine hydrochloride
Storage condition: Sealed in dry, Room Temperature
Formula:
C₈H₁₀ClF₂N Storage Temperature: Ambient |
MDL Number:
MFCD11101244 CAS Number: 844647-37-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...