8-Methyl-4-quinazolone 97%

BD216451-250MG BD216451-1G
BLDPBD216451-250MGEA 17.9 EUR
BLDPBD216451-250MG BLDPBD216451-1G
8-Methyl-4-quinazolone 97%
8-Methyl-4-quinazolone

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD01685945
CAS Number: 19181-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR