(2S,3S)-(+)-2,3-Butanediol 97%
|
|
BD2165-25G
BD2165-5G
BLDPBD2165-25GEA
553
EUR
BLDPBD2165-25G
BLDPBD2165-5G
(2S,3S)-(+)-2,3-Butanediol 97%
(2S,3S)-(+)-2,3-Butanediol
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₄H₁₀O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00063648 CAS Number: 19132-06-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...