4-Bromo-2-methylbenzyl alcohol 98%
|
|
BD216810-5G
BD216810-250MG
BLDPBD216810-5GEA
86.1
EUR
BLDPBD216810-5G
BLDPBD216810-250MG
4-Bromo-2-methylbenzyl alcohol 98%
4-Bromo-2-methylbenzyl alcohol
Storage condition: Sealed in dry, Room Temperature
Formula:
C₈H₉BrO MW: 201.06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11847398 CAS Number: 17100-58-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...