2-Bromo-1,3-dimethyl-5-fluorobenzene 98%

BD216873-1G BD216873-100G
BLDPBD216873-1GEA 9.5 EUR
BLDPBD216873-1G BLDPBD216873-100G
2-Bromo-1,3-dimethyl-5-fluorobenzene 98%
2-Bromo-1,3-dimethyl-5-fluorobenzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₈BrF
Storage Temperature: Ambient
MDL Number: MFCD09864702
CAS Number: 14659-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR